聚丙烯公凸轮式 NPT 公螺纹转接头
供应商: MASTERFLEX PREMIUM
34020-02
MFLB34020-02EA 0 CNY
MFLB34020-02
聚丙烯公凸轮式 NPT 公螺纹转接头
接头
Cam couplers provide quick disconnection from system.
  • All fittings have EPDM gaskets
  • Constructed of durable polypropylene
Cam couplers are ideal for providing a quick-disconnect solution to your pump line.