Masterflex® Low Pressure Drop Gas Mass Flowmeter for Gases, Avantor®
供应商: MASTERFLEX PREMIUM
32935-09 32935-11 32935-27 32935-31 32935-32 32935-37 32935-48 32935-00 32935-01 32935-04 32935-06 32935-08 32935-10 32935-12 32935-13 32935-14 32935-16 32935-17 32935-18 32935-19 32935-20 32935-21 32935-22 32935-23 32935-26 32935-28 32935-29 32935-30 32935-33 32935-34 32935-35 32935-38 32935-39 32935-40 32935-42 32935-43 32935-44 32935-46 32935-47 32935-49 32935-50 32935-51 32935-52 32935-54 32935-56 32935-58 32935-59
MFLB32935-09EA 0 CNY
MFLB32935-09 MFLB32935-11 MFLB32935-27 MFLB32935-31 MFLB32935-32 MFLB32935-37 MFLB32935-48 MFLB32935-00 MFLB32935-01 MFLB32935-04 MFLB32935-06 MFLB32935-08 MFLB32935-10 MFLB32935-12 MFLB32935-13 MFLB32935-14 MFLB32935-16 MFLB32935-17 MFLB32935-18 MFLB32935-19 MFLB32935-20 MFLB32935-21 MFLB32935-22 MFLB32935-23 MFLB32935-26 MFLB32935-28 MFLB32935-29 MFLB32935-30 MFLB32935-33 MFLB32935-34 MFLB32935-35 MFLB32935-38 MFLB32935-39 MFLB32935-40 MFLB32935-42 MFLB32935-43 MFLB32935-44 MFLB32935-46 MFLB32935-47 MFLB32935-49 MFLB32935-50 MFLB32935-51 MFLB32935-52 MFLB32935-54 MFLB32935-56 MFLB32935-58 MFLB32935-59
Masterflex® Low Pressure Drop Gas Mass Flowmeter for Gases, Avantor®
Flow Switches
Achieve high accuracy and low pressure drop!
  • Full-scale pressure drops as low as 0.1 psid to 0.69 psid
  • Measure 30 standard gases with a 200:1 turndown ratio
  • Set up and control from an easy-to-use keypad
Using a newer laminar differential pressure flow technology, these mass flow meters and controllers have a much lower pressure drop across the instrument. Meters and controllers require no warm-up time, and offer a 10 ms response time for meters and 100 ms for controllers. Typical applications include air sampling, natural gas metering and calibration,More Product Information