Oxygen (O2) Gas Mass Flow Controllers with Metal bodies
供应商: MASTERFLEX PREMIUM
32660-32 32660-34 32660-42 32660-48
MFLB32660-32EA 0 CNY
MFLB32660-32 MFLB32660-34 MFLB32660-42 MFLB32660-48
Oxygen (O2) Gas Mass Flow Controllers with Metal bodies
Flow Switches
Precisely control flow rates with built-in valve.
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These controllers feature an advanced straight-tube sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity.
The flow rate set point can be established byMore Product Information