PrimeLock® Installation Tools for High-Purity PFA Fittings
供应商: MASTERFLEX PREMIUM
30493-25
MFLB30493-25EA 0 CNY
MFLB30493-25
PrimeLock® Installation Tools for High-Purity PFA Fittings
工具套件
Easily install your High-Purity PFA Fittings.
  • Optimize performance of your PFA PrimeLock® Fittings
Handheld and benchtop installation tools help you to quickly and easily install the PrimeLock insert correctly every time. Benchtop tool is the quickest and easiest method of installing the insert when space constraints are not an issue. The compact, handheld tool enables you to install the insert in remote locations. Installation tools require mandrel(s) and clamp insert(s) for operation.