GF Signet Metal Flowmeter Installation Tee Fittings
供应商: MASTERFLEX PREMIUM
05615-26 05615-27 05620-41 05620-43 05620-45 05620-46 05620-56
MFLB05615-26EA 0 CNY
MFLB05615-26 MFLB05615-27 MFLB05620-41 MFLB05620-43 MFLB05620-45 MFLB05620-46 MFLB05620-56
GF Signet Metal Flowmeter Installation Tee Fittings
流量计
Ensure proper insertion depth and measurement.
  • Choose from 316 Stainless Steel (SS), carbon steel (CS), galvanized iron (schedule 40), and copper tee fittings
These metal installation tee fittings are available in 316 Stainless Steel (SS), carbon steel (CS), galvanized iron (schedule 40), and copper materials in a wide range of sizes from 3/4” to 8” pipe inner diameters.
Standard metal tees constructed of SS, CS, and galvanized iron fittings include PVDF insert for all sizes and NPT(F) threads at each end. Copper tee fittings include PVDF insert forMore Product Information