Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® S3™ E-Lab, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA96461-09EA 0 CNY
MFLA96461-09 MFLA96461-10 MFLA96461-12 MFLA96461-14 MFLA96461-16 MFLA96461-18 MFLA96461-19 MFLA96461-22 MFLA96461-24 MFLA96461-26 MFLA96461-27 MFLA96461-28 MFLA96461-29 MFLA96461-30 MFLA96461-31 MFLA96461-32 MFLA96461-34 MFLA96461-36 MFLA96461-38 MFLA96461-39 MFLA96461-40 MFLA96461-42 MFLA96461-44 MFLA96461-46 MFLA96461-48 MFLA96461-49
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® S3™ E-Lab, Avantor®
胶管 泵管
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Ideal for general laboratory applications
  • Clear for easy sample viewing
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes inMore Product Information