Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PFL Single Elements, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA96252-00EA 0 CNY
MFLA96252-00 MFLA96252-02 MFLA96252-06 MFLA96252-10 MFLA96252-12 MFLA96252-16 MFLA96252-18 MFLA96252-24 MFLA96252-26 MFLA96252-30 MFLA96252-34 MFLA96252-40 MFLA96252-04 MFLA96252-08 MFLA96252-22 MFLA96252-28 MFLA96252-38
Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PFL Single Elements, Avantor®
胶管 泵管
Excellent choice for high pressure, low spallation peristaltic tubing applications.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Engineered for long life in peristaltic pump applications
  • Unique composite of expanded PTFE and platinum-cured perfluoroelastomer
Applications: Chemical, criticalMore Product Information