Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PCS Single Elements, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA96251-02EA 0 CNY
MFLA96251-02 MFLA96251-06 MFLA96251-08 MFLA96251-16 MFLA96251-26 MFLA96251-28 MFLA96251-30 MFLA96251-34 MFLA96251-40 MFLA96251-42 MFLA96251-58 MFLA96251-62 MFLA96251-04 MFLA96251-10 MFLA96251-12 MFLA96251-20 MFLA96251-24 MFLA96251-32 MFLA96251-36 MFLA96251-48 MFLA96251-50 MFLA96251-52
Masterflex® Ismatec® Pump Tubing, GORE® STA-PURE® Series PCS Single Elements, Avantor®
胶管 泵管
Excellent choice for high pressure, low spallation peristaltic tubing applications.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Engineered for long life in peristaltic pump applications
  • Unique composite of expanded PTFE and platinum-cured silicone
Applications: Chemical, criticalMore Product Information