Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® Fuel and Lubricant, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA95703-18EA 0 CNY
MFLA95703-18 MFLA95703-32 MFLA95703-14 MFLA95703-22 MFLA95703-24 MFLA95703-26 MFLA95703-48 MFLA95703-36
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® Fuel and Lubricant, Avantor®
胶管 泵管
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Specially designed to transport hydrocarbons, petroleum products, or distallates
  • Transparent yellow color
Appearance: Opaque Temperature: -70 to 52 °C Life at 100 rpm: 1000 hrs Advantages:LongMore Product Information