Masterflex® Ismatec® Reglo Digital Miniflex Pumps, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA78018-40EA 0 CNY
MFLA78018-40 MFLA78018-42 MFLA78018-40-AU MFLA78018-42-AU MFLA78018-40-DK MFLA78018-42-DK MFLA78018-40-IN MFLA78018-42-IN MFLA78018-40-USS MFLA78018-42-USS MFLA78018-40-IL MFLA78018-42-IL MFLA78018-40-EU MFLA78018-42-EU MFLA78018-40-UK MFLA78018-42-UK MFLA78018-40-CH MFLA78018-42-CH MFLA78018-40-IT MFLA78018-42-IT
Masterflex® Ismatec® Reglo Digital Miniflex Pumps, Avantor®
蠕动泵
Versatile pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Anti-drip and speed ramping
More Product Information