Masterflex® Ismatec® Reglo Digital Multichannel Pumps, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA78018-10EA 0 CNY
MFLA78018-10 MFLA78018-12 MFLA78018-14 MFLA78018-20 MFLA78018-22 MFLA78018-24 MFLA78018-10-AU MFLA78018-12-AU MFLA78018-14-AU MFLA78018-20-AU MFLA78018-22-AU MFLA78018-24-AU MFLA78018-10-DK MFLA78018-12-DK MFLA78018-14-DK MFLA78018-20-DK MFLA78018-22-DK MFLA78018-24-DK MFLA78018-10-IN MFLA78018-12-IN MFLA78018-14-IN MFLA78018-20-IN MFLA78018-22-IN MFLA78018-24-IN MFLA78018-10-USS MFLA78018-12-USS MFLA78018-14-USS MFLA78018-20-USS MFLA78018-22-USS MFLA78018-24-USS MFLA78018-10-IL MFLA78018-12-IL MFLA78018-14-IL MFLA78018-20-IL MFLA78018-22-IL MFLA78018-24-IL MFLA78018-10-EU MFLA78018-12-EU MFLA78018-14-EU MFLA78018-20-EU MFLA78018-22-EU MFLA78018-24-EU MFLA78018-10-UK MFLA78018-12-UK MFLA78018-14-UK MFLA78018-20-UK MFLA78018-22-UK MFLA78018-24-UK MFLA78018-10-CH MFLA78018-12-CH MFLA78018-14-CH MFLA78018-20-CH MFLA78018-22-CH MFLA78018-24-CH MFLA78018-10-IT MFLA78018-12-IT MFLA78018-14-IT MFLA78018-20-IT MFLA78018-22-IT MFLA78018-24-IT
Masterflex® Ismatec® Reglo Digital Multichannel Pumps, Avantor®
蠕动泵
Versatile multichannel pumps for accurate low-flow fluid delivery – with an intuitive touch-screen interface.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Anti-drip and speed
More Product Information