Masterflex® I/P®精密无刷工艺泵,Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA77963-10EA 0 CNY
MFLA77963-10 MFLA77963-20
Masterflex® I/P®精密无刷工艺泵,Avantor®
蠕动泵
Powerful, low-maintenance drives are versatile and cost-effective.
  • Digital interface with LED display shows percent speed with 1% resolution
  • Speed control via three-turn potentiometer for quick, easy adjustments
  • Easily control start/stop and reverse features via sealed keypad
  • Contructed of non-chip painted steel and aluminum IP55-rated housing
  • Brushless, maintenance-free motor offers ±0.25% speed control accuracy
I/P® brushless process drives offer heavy-duty performance for your demanding process applications. This model features robust 1⁄3-hpMore Product Information