Masterflex® L/S® Network-Compatible Pump Systems, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA77975-60EA 0 CNY
MFLA77975-60 MFLA77924-70-USS MFLA77924-82-USS MFLA77975-60-USS MFLA77975-62-USS MFLA77975-65-USS MFLA77975-77-USS
Masterflex® L/S® Network-Compatible Pump Systems, Avantor®
蠕动泵
Networking drives offer either native EtherNet/IP™ or Profibus communication—no additional hardware or firmware required.
  • Flow rate 0.001 to 3400 ml/min with L/S tubing—flow rate depends on drive rpm pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—
More Product Information