Masterflex® Ismatec® Reglo Miniflex® Dispensing Pump Systems, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA77922-22EA 0 CNY
MFLA77922-22 MFLA77922-22-AU MFLA77922-22-DK MFLA77922-22-IN MFLA77922-22-USS MFLA77922-22-IL MFLA77922-22-EU MFLA77922-22-UK MFLA77922-22-CH MFLA77922-22-IT
Masterflex® Ismatec® Reglo Miniflex® Dispensing Pump Systems, Avantor®
蠕动泵
Complete systems to increase the efficiency and accuracy of filling and dispensing applications.
  • Flow range 0.16 to 365 ml/min with included L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Realize increased efficiency and throughput when compared to hand pouring, manually pipetting, or using bottletop style dispensers
  • 5” (12.7 cm) capacitive TFT/RGB touch screen puts the intuitive interface at your fingertips
  • Dispense by volume or time -- anti-drip and speed ramping available in Volume dispense mode
  • Flexible operational control via
More Product Information