Masterflex® L/S® MasterSense™ Pump Systems, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA77920-42EA 0 CNY
MFLA77920-42 MFLA77920-42-AU MFLA77920-42-DK MFLA77920-42-IN MFLA77920-42-USS MFLA77920-42-IL MFLA77920-42-EU MFLA77920-42-UK MFLA77920-42-CH MFLA77920-42-IT MFLA77920-50 MFLA77976-00 MFLA77976-02 MFLA77976-10 MFLA77976-12 MFLA77976-00-AU MFLA77976-02-AU MFLA77976-10-AU MFLA77976-12-AU MFLA77920-50-AU MFLA77920-40 MFLA77920-52
Masterflex® L/S® MasterSense™ Pump Systems, Avantor®
蠕动泵
Complete pump systems for accurate fluid delivery - with an intuitive touch-screen interface.
  • Precise, repeatable performance that is quick and easy to set up and program
  • 5" (12,7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Over-the-air firmware updates let you take full advantage of improvements in drive features and functionality
  • Options for connectivity and monitoring include MasterflexLive, EtherNet/IP™, Profibus, or analog signal
More Product Information