Masterflex® L/S® Variable-Speed Economy Modular Drive Systems, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA77913-02EA 0 CNY
MFLA77913-02 MFLA77913-07 MFLA77913-12 MFLA77913-17 MFLA77913-07-AU MFLA77913-17-AU MFLA77913-07-DK MFLA77913-17-DK MFLA77913-07-IN MFLA77913-17-IN MFLA77913-07-IL MFLA77913-17-IL MFLA77913-07-EU MFLA77913-17-EU MFLA77913-07-UK MFLA77913-17-UK MFLA77913-07-CH MFLA77913-17-CH MFLA77913-07-IT MFLA77913-17-IT
Masterflex® L/S® Variable-Speed Economy Modular Drive Systems, Avantor®
蠕动泵
Versatile variable-speed pump system with a wide flow range and simple controls.
  • Separate motor and controller are connected via a 6-ft (1.9 m) cable — place components where convenient or where space allows
  • Single-turn speed control and on/off switch with green power indicator — maintain speed setting while turning pump on/off
  • The 1/10-hp reversible motor has soft start and back EMF for ±2% speed control
  • Painted steel housing is IP33 rated for protection from vertically falling water
Choose a convenient, complete system all with one catalog number. AMore Product Information