Masterflex® L/S® High-Performance Pump Heads, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA77252-62EA 0 CNY
MFLA77252-62 MFLA77252-70 MFLA77252-72 MFLA77252-80 MFLA77252-82 MFLA77252-60
Masterflex® L/S® High-Performance Pump Heads, Avantor®
Pump Heads Peristaltic Pump Heads
All-new patented design offers faster tubing changes and improved ergonomics.
  • Tubing occlusion is automatic and optimized when pump head is closed and latched – load tubing in 15 seconds or less
  • Flow rates up to 3400 ml/min—flow rate depends on drive rpm, pump head, and tubing size
  • Pressure up to 150 psi (10.3 bar) with L/S high-pressure (HP) pump tubing
  • Reinforced nylon housing and occlusion bed with stainless steel rotor/roller assembly
  • Compatible with all 1/10 hp L/S drives that accept two or more pump heads
  • Compact design for efficient use of space
More Product Information