Masterflex® B/T® MasterSense™ BioPharma Process Pump, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA77112-10EA 0 CNY
MFLA77112-10 MFLA77112-10-AU MFLA77112-10-DK MFLA77112-10-IN MFLA77112-10-USS MFLA77112-10-IL MFLA77112-10-EU MFLA77112-10-UK MFLA77112-10-CH MFLA77112-10-IT
Masterflex® B/T® MasterSense™ BioPharma Process Pump, Avantor®
蠕动泵
High-flow pump for process applications – with an intuitive touch-screen interface and advanced connectivity.
  • Powerful pump engineered for continuous-duty biopharma process applications
  • 316 stainless steel housing is IP66 rated for use in washdown environments
  • 5" (12,7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Adjust parameters with a simple touch, even while wearing gloves
  • Over-the-air firmware updates let you take full advantage of future improvements in pump features and functionality
  • Options for connectivity and
More Product Information