Masterflex® EtherNet/IP™ Compatible Drives Powering Quattroflow™ Pumps
供应商: MASTERFLEX PROPRIETARY
MFLA74000-49EA 0 CNY
MFLA74000-49 MFLA74000-51 MFLA74000-53 MFLA74000-59 MFLA74000-49-AU MFLA74000-51-AU MFLA74000-53-AU MFLA74000-59-AU MFLA74000-49-DK MFLA74000-51-DK MFLA74000-53-DK MFLA74000-59-DK MFLA74000-49-IN MFLA74000-51-IN MFLA74000-53-IN MFLA74000-59-IN MFLA74000-49-USS MFLA74000-51-USS MFLA74000-53-USS MFLA74000-59-USS MFLA74000-49-IL MFLA74000-51-IL MFLA74000-53-IL MFLA74000-59-IL MFLA74000-49-EU MFLA74000-51-EU MFLA74000-53-EU MFLA74000-59-EU MFLA74000-49-UK MFLA74000-51-UK MFLA74000-53-UK MFLA74000-59-UK MFLA74000-49-CH MFLA74000-51-CH MFLA74000-53-CH MFLA74000-59-CH MFLA74000-49-IT MFLA74000-51-IT MFLA74000-53-IT MFLA74000-59-IT
Masterflex® EtherNet/IP™ Compatible Drives Powering Quattroflow™ Pumps
Diaphragm Pumps
Monitor transfer of delicate biological fluids with EtherNet/IP™ control capability.
  • Choose from readily available single-use or multiple-use Quattroflow™ pump head systems
  • Advanced network and communication models feature EtherNet/IP™
  • Diaphragms enable soft, gentle pumping through the strokes generated by the eccentric shaft
  • No mechanical seal or rotating parts that can fail—only the diaphragm and body come in contact with the fluid
  • Ideal for cross-flow systems, tangential flow filtration (TFF), chromatography, cultures, and more
The Quattroflow™More Product Information