Value Plastics® Adapter Fittings, Male Luer to Hose Barb, Straight, Polycarbonate
供应商: MASTERFLEX PROPRIETARY
MFLA45504-50BLKEA 0 CNY
MFLA45504-50BLK MFLA45504-08BLK MFLA45504-00BLK MFLA45504-06BLK MFLA45504-02BLK MFLA45504-07BLK MFLA45504-19BLK MFLA45504-04BLK
Value Plastics® Adapter Fittings, Male Luer to Hose Barb, Straight, Polycarbonate
接头
Ensure your process integrity with full material traceability.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly
More Product Information