Masterflex® Adapter Fittings, Luer to Threaded, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA45502-60EA 0 CNY
MFLA45502-60 MFLA45502-66 MFLA45505-76 MFLA45505-78 MFLA45505-80 MFLA45505-82 MFLA45505-84 MFLA45505-86 MFLA45505-88
Masterflex® Adapter Fittings, Luer to Threaded, Avantor®
接头
Easily transition to a luer fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs,
More Product Information