Masterflex® Male Rotating Luer Lock Plug Fitting, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA40320-00EA 0 CNY
MFLA40320-00 MFLA40320-01 MFLA40320-02 MFLA40320-03 MFLA40320-04 MFLA40320-05 MFLA40320-06 MFLA40320-07 MFLA40320-08 MFLA40320-09 MFLA40320-10 MFLA40320-11 MFLA40320-12 MFLA40320-13 MFLA40320-14 MFLA40320-15 MFLA40320-16 MFLA40320-17 MFLA40320-18 MFLA40320-19 MFLA40320-20
Masterflex® Male Rotating Luer Lock Plug Fitting, Avantor®
接头
Easily transition to a luer fitting.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Determine which connector type is needed for your application by contacting free technical application assistance
These fittings are manufactured in an ISO Class 7 cleanroom at an ISO 9001 facility; utilizing an ISO 13485 quality system. There is full lot traceability and tight dimensional tolerances forMore Product Information