Masterflex® Single-Use Ultrasonic Flow Sensors, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA33501-16EA 0 CNY
MFLA33501-16 MFLA33501-17 MFLA33501-36 MFLA33501-73 MFLA33501-82 MFLA33501-15 MFLA33501-24 MFLA33501-25 MFLA33501-91
Masterflex® Single-Use Ultrasonic Flow Sensors, Avantor®
Fluid Sensors
Obtain accurate flow readings without introducing a new material to the fluid path.
  • Noninvasive design eliminates contact with working fluid
  • Ideally suited for strict hygienic standards
  • Continuously monitor peristaltic pump flow rates and send data directly to your computer
  • Compact aluminum sensor body clamps around the tubing
  • Integrated electronics—no external electronic board is needed
  • Specifically calibrated for use with Masterflex L/S® and I/P® platinum-cured silicone tubing
Masterflex® ultrasonic flow sensors accurately and noninvasively measureMore Product Information