Masterflex® 65毫米直接读数面板安装流量计,Avantor®
供应商: MASTERFLEX PROPRIETARY
32003-09 32004-09 32005-09 32013-01 32013-16 32013-18 32013-21 32013-24 32013-28 32013-29 32013-31 32013-35 32014-01 32014-07 32014-11 32014-15 32014-16 32014-21 32014-28 32014-29 32015-24 32015-33
MFLA32013-03EA 0 CNY
MFLA32013-03 MFLA32013-05 MFLA32013-07 MFLA32013-09 MFLA32013-11 MFLA32013-13 MFLA32013-15 MFLA32013-17 MFLA32013-19 MFLA32013-27 MFLA32013-33 MFLA32014-03 MFLA32014-05 MFLA32014-09 MFLA32014-13 MFLA32014-17 MFLA32014-24 MFLA32015-01 MFLA32015-03 MFLA32015-05 MFLA32015-07 MFLA32015-09 MFLA32015-11 MFLA32015-13 MFLA32015-15 MFLA32015-16 MFLA32015-17 MFLA32015-19 MFLA32015-21 MFLA32015-23 MFLA32015-29 MFLA32015-31 MFLA32015-35 MFLB32003-09 MFLB32004-09 MFLB32005-09 MFLB32013-01 MFLB32013-16 MFLB32013-18 MFLB32013-21 MFLB32013-24 MFLB32013-28 MFLB32013-29 MFLB32013-31 MFLB32013-35 MFLB32014-01 MFLB32014-07 MFLB32014-11 MFLB32014-15 MFLB32014-16 MFLB32014-21 MFLB32014-28 MFLB32014-29 MFLB32015-24 MFLB32015-33
Masterflex® 65毫米直接读数面板安装流量计,Avantor®
流量计
No more eye strain - front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
More Product Information