Masterflex® Tuf-Steel® Sanitary Screen Tri-Clamp Gaskets, Avantor®
供应商: MASTERFLEX PROPRIETARY
30539-20 30539-40 30539-44 30539-48 30539-68 30539-84 30539-86 30539-88
MFLA30539-08EA 0 CNY
MFLA30539-08 MFLA30539-26 MFLA30539-28 MFLA30539-46 MFLA30539-66 MFLB30539-20 MFLB30539-40 MFLB30539-44 MFLB30539-48 MFLB30539-68 MFLB30539-84 MFLB30539-86 MFLB30539-88
Masterflex® Tuf-Steel® Sanitary Screen Tri-Clamp Gaskets, Avantor®
密封件 Gaskets
Protect your pump, valves and other components in fluid line.
  • Pre-filter before critical membrane filters
  • Generate turbulence for enhanced mass or heat transfer
  • Purify in chromatography applications
Screen gaskets are designed to be interchangeable with standard sanitary clamp gaskets. Gaskets are available in platinum-cured silicone, Viton®, EPDM, PTFE, and Tuf-Steel in mesh sizes of 10, 20, 40, 60 and 100. Screen mesh is constructed of 316L SS. Gaskets meet USP Class VI, USDA, 3-A and cGMP criteria. FDA compliant. All gaskets are animal-derivedMore Product Information