Masterflex® L/S® Precision Process Drives, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA07528-40EA 0 CNY
MFLA07528-40 MFLA07528-50 MFLA07528-40-AU MFLA07528-50-AU MFLA07528-40-DK MFLA07528-50-DK MFLA07528-40-IN MFLA07528-50-IN MFLA07528-40-USS MFLA07528-45-USS MFLA07528-50-USS MFLA07528-40-IL MFLA07528-50-IL MFLA07528-40-EU MFLA07528-50-EU MFLA07528-40-UK MFLA07528-50-UK MFLA07528-40-CH MFLA07528-50-CH MFLA07528-40-IT MFLA07528-50-IT
Masterflex® L/S® Precision Process Drives, Avantor®
Pump Drives Peristaltic Pump Drives
Easy-to-use drives offer precise speed control and digital display of rpm—NEMA 4X housings stand up to process and washdown applications.
  • Flow rate 0.06 to 3400 ml/min (depends on drive rpm and tubing size)
  • Four digit LED display shows motor rpm—confirm speed setting at a glance
  • Integrated open-head sensor stops drive motor when pump head is opened
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Powerful motors let you stack 2 or 4 pump heads for increased flow rates and for
More Product Information