Masterflex® MasterSense™ MP1 PendoTECH™ Pressure Sensor Adaptor Kits, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA07526-90EA 0 CNY
MFLA07526-90 MFLA07526-91 MFLA07526-92
Masterflex® MasterSense™ MP1 PendoTECH™ Pressure Sensor Adaptor Kits, Avantor®
蠕动泵
Designed for simplified integration of a single PendoTECH™ pressure sensor into Masterflex® MasterSense™ pump systems.
  • Add critical pressure monitoring to your workflow
  • Specifically designed for PendoTECH™ single-use pressure sensors
  • Perfect for tangential-flow filtration, depth filtration, harvest, bulk transfer, chromatography and fill applications
  • IP 66 washdown housing
  • CE compliance
  • ABS housing for wipe-down and cleanability
asterSense™ M1 adaptors allow for simplified connection of PendoTECH™ single-use pressure sensors into MasterSense™More Product Information