Masterflex® L/S® MasterSense™ Drives, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA07526-10EA 0 CNY
MFLA07526-10 MFLA07526-12 MFLA07526-20 MFLA07526-10-AU MFLA07526-12-AU MFLA07526-20-AU MFLA07526-10-DK MFLA07526-12-DK MFLA07526-20-DK MFLA07526-10-IN MFLA07526-12-IN MFLA07526-20-IN MFLA07526-10-USS MFLA07526-12-USS MFLA07526-20-USS MFLA07526-10-IL MFLA07526-12-IL MFLA07526-20-IL MFLA07526-10-EU MFLA07526-12-EU MFLA07526-20-EU MFLA07526-10-UK MFLA07526-12-UK MFLA07526-20-UK MFLA07526-10-CH MFLA07526-12-CH MFLA07526-20-CH MFLA07526-10-IT MFLA07526-12-IT MFLA07526-20-IT MFLA07526-22 MFLA07526-22-AU
Masterflex® L/S® MasterSense™ Drives, Avantor®
Pump Drives Peristaltic Pump Drives
Versatile pumps for accurate fluid delivery – with an intuitive touch-screen interface.
  • Powerful, space-efficient drives will accept all L/S pump head and tubing options
  • 5" (12,7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Over-the-air firmware updates let you take full advantage of improvements in drive features and functionality
  • Options for connectivity and monitoring include MasterflexLive, EtherNet/IP™, Profibus, or analog signal
  • Three-
More Product Information