Masterflex® L/S® Easy-Load® Pump Heads for High-Performance Precision Tubing, Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA07514-12EA 0 CNY
MFLA07514-12 MFLA07514-32
Masterflex® L/S® Easy-Load® Pump Heads for High-Performance Precision Tubing, Avantor®
Pump Heads Peristaltic Pump Heads
Save time with fast tubing changes and reduced maintenance.
  • New design and automatic tubing retention allow for multiple tubing sizes—change tubing without removing head from drive
  • Rotor is constructed of stainless steel (SS) for enhanced durability
  • Pump heads are stackable for multichannel applications
  • Compact design for efficient use of space
  • Suitable for use with L/S® High-Performance Precision pump tubing sizes
This Easy-Load® pump head accepts high-performance precision tubing— ensures optimal performance from your Masterflex pump. Select a pumpMore Product Information