Masterflex® PTFE 泵管,Avantor®
供应商: MASTERFLEX PROPRIETARY
MFLA06407-41EA 0 CNY
MFLA06407-41 MFLA06605-27 MFLA06605-28 MFLA06605-29 MFLA06605-30 MFLA06605-31 MFLA06605-32 MFLA06605-33 MFLA06605-34 MFLA06605-35 MFLA06605-36 MFLA06605-37 MFLA06605-38 MFLA06605-39 MFLA06605-40 MFLA06605-41 MFLA06605-42 MFLA06605-44 MFLA06605-45 MFLA06605-46 MFLA06605-48 MFLA06605-49 MFLA06605-50 MFLA06605-51
Masterflex® PTFE 泵管,Avantor®
胶管 Transfer Tubing
Won't crack under heat or pressure.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Demanding lab, chemical, electrical, mechanical, pharmaceutical, and automotive applications. Good for high-purity work. PTFE tubing withstands extreme temperatures.
Characteristics: Chemically inert, nontoxic, andMore Product Information