Masterflex® 装有高分辨率阀门的校正流量计,Avantor®
供应商: MASTERFLEX PROPRIETARY
03217-58 03217-63 03217-68 03217-74 03217-76 03217-79 03227-04 03227-08 03227-12 03227-14 03227-20 03227-24 03227-84 03229-05 03229-09 03229-13 03229-25 03295-04 03295-06 03295-08 03295-12 03295-14 03295-20 03295-22 03295-24 03295-26 03295-28
MFLA03217-53EA 0 CNY
MFLA03217-53 MFLA03217-57 MFLA03217-62 MFLA03217-67 MFLA03217-72 MFLA03217-73 MFLA03227-06 MFLA03227-10 MFLA03227-16 MFLA03227-18 MFLA03227-22 MFLA03227-26 MFLA03227-28 MFLA03227-30 MFLA03229-07 MFLA03229-11 MFLA03229-15 MFLA03229-17 MFLA03229-19 MFLA03229-21 MFLA03229-23 MFLA03229-27 MFLA03229-29 MFLA03229-31 MFLA03295-10 MFLA03295-16 MFLA03295-18 MFLB03217-58 MFLB03217-63 MFLB03217-68 MFLB03217-74 MFLB03217-76 MFLB03217-79 MFLB03227-04 MFLB03227-08 MFLB03227-12 MFLB03227-14 MFLB03227-20 MFLB03227-24 MFLB03227-84 MFLB03229-05 MFLB03229-09 MFLB03229-13 MFLB03229-25 MFLB03295-04 MFLB03295-06 MFLB03295-08 MFLB03295-12 MFLB03295-14 MFLB03295-20 MFLB03295-22 MFLB03295-24 MFLB03295-26 MFLB03295-28
Masterflex® 装有高分辨率阀门的校正流量计,Avantor®
流量计
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet,More Product Information