Reference electrode filling solutions
供应商: Mettler - Toledo
51343182 51343180 51350074 51350072 51350082
METT51343182EA 0 CNY
METT51343182 METT51343180 METT51350074 METT51350072 METT51350082
Reference electrode filling solutions
电解质 电极填充溶液
FRISCOLYT-B® (84,4% glycerol, 11% KCl and 4,6% H₂O) is for low temperature samples.