Shelving systems, stem castor carts with open grid shelves, MetroMax Q®
供应商: METRO
Q536EG3 Q556EG3 Q566BG3 Q356EG3 Q336EG3 Q436EG3
METOQ536EG3EA 0 CNY
METOQ536EG3 METOQ556EG3 METOQ566BG3 METOQ356EG3 METOQ336EG3 METOQ436EG3
Shelving systems, stem castor carts with open grid shelves, MetroMax Q®
家具 货架
Corrosion proof shelves and posts with removable polymer mats.
  • Load capacity: 408 kg (for models with PU castors); 272 kg (for models with resilient rubber castors)
Standard unit consist of 5 shelves (4 open grid, 1 bottom solid shelf), posts, wedge connectors and choice of PU or resilient rubber castors.