Transport trolleys for glassware, MetroMax i®
供应商: METRO
GWBX3-1 GWBX3-2 GWTX3-2 GWTX3-1
METOGWBX3-1EA 0 CNY
METOGWBX3-1 METOGWBX3-2 METOGWTX3-2 METOGWTX3-1
Transport trolleys for glassware, MetroMax i®
手推车
MetroMax i® glassware carts are available in two and four basket capacities and are ideal for transporting glassware safely in the laboratory.
  • The solid bottom shelf provides spill retention and can be removed from shelf frame for easy cleaning
订购须知 Castors must be ordered separately.