Shelving starter and add-on units, MetroMax Q®
供应商: METRO
5AQ557G3 5Q337G3 5Q357G3 5Q367G3 5Q537G3 5Q557G3 5Q567G3 5AQ357G3 5AQ457G3 5AQ467G3 5Q457G3 5Q467G3 5AQ337G3 5Q347G3 5Q437G3 5AQ567G3 5Q577G3
METO5AQ557G3EA 0 CNY
METO5AQ557G3 METO5Q337G3 METO5Q357G3 METO5Q367G3 METO5Q537G3 METO5Q557G3 METO5Q567G3 METO5AQ357G3 METO5AQ457G3 METO5AQ467G3 METO5Q457G3 METO5Q467G3 METO5AQ337G3 METO5Q347G3 METO5Q437G3 METO5AQ567G3 METO5Q577G3
Shelving starter and add-on units, MetroMax Q®
家具 货架
These Super Adjustable 2™ Starter and Add-On units save time and money by eliminating adjacent posts and replacing them with 'S' hooks. This system offers a convenient way to create long runs of shelving, shelving at right angles and combinations of runs and right angles.
The load capacity evenly distributed for shelf lengths up to 1219 mm is 363 kg per shelf and for shelf lengths 1524 mm and longer is 272 kg per shelf.
Chrome plated starter units includes four 1880 mm post and five 457 mm deep Super Adjustable 2™ shelves. Add-on units includes two 1880 mm post and five 457 mm deep SuperMore Product Information