Semi-micro and analytical balances, NewClassic series
供应商: METTLER TOLEDO
11124474 11124477 11124480 11142062 11142056 11142068
METB11124474EA 0 CNY
METB11124474 METB11124477 METB11124480 METB11142062 METB11142056 METB11142068
Semi-micro and analytical balances, NewClassic series
天平
A stylish series of traditional analytical balances for simple weighing tasks. The balances all include the following benefits with more specific features listed for the three ranges below:
  • SmartTrac; shows how much of the balance range has been used
  • Balance diagnostics
  • Protection of balance configurations against manipulation and also overload protection
Semi-micro NewClassic MS balances feature:
HRT (HiRes Technology) weighing cell
FACT fully automatic time and temperature controlled internal adjustment
ErgoDoor; the right door of the draught shieldMore Product Information