Precision balances, Excellence Plus XP series
供应商: METTLER TOLEDO
11130022 11130020 11130040 11130000 11130070 11130102 11130100 11130101 11130201 11130200 11130206 11130141 11130140 11130221 11130220 11130088 11130094 11130039 11130060 11130061
METB11130022EA 0 CNY
METB11130022 METB11130020 METB11130040 METB11130000 METB11130070 METB11130102 METB11130100 METB11130101 METB11130201 METB11130200 METB11130206 METB11130141 METB11130140 METB11130221 METB11130220 METB11130088 METB11130094 METB11130039 METB11130060 METB11130061
Precision balances, Excellence Plus XP series
天平
The XP precision balances provide high productivity instruments with improved linearity, safety and security using MonoBloc HighSpeed technology. This is achieved with the introduction of QM toolbox, SmartSens and MinWeigh functions. This range is therefore particularly suited where regulatory compliance is required. Draught shields are standard on the small square platform size (see models denoted * in the table). The touch screen provides fast and easy access to all balance functions.
  • Colour-coded user profiles for up to eight password protected users
  • SmartSens provides hands-free
More Product Information