Industrial scales, NewClassic, MS series
供应商: METTLER TOLEDO
11124573 11124576 11124588 11124704 11124671 11124683 11124701
METB11124573EA 0 CNY
METB11124573 METB11124576 METB11124588 METB11124704 METB11124671 METB11124683 METB11124701
Industrial scales, NewClassic, MS series
天平
A familiar, attractive design of large pan scales that incorporates new technology to deliver rugged, reliable, effective performance. Applications include formulation, statistics, checkweighing, piece counting.
  • MonoBloc weighing technology for fast, repeatable results
  • High grade chemically resistant die-cast aluminium housing
  • USB connectivity to PC for easy transfer of results (on models except KLIPE)
  • Diagnostics check helps audit the balance adjustment history
  • One-touch function via user-defined SmartKeys
订购须知 OIML approved versions (denoted /M)More Product Information