Semi-micro and analytical balances, Excellence XA and XS series
供应商: METTLER TOLEDO
11106009 11106000 11106015 11106003 11106006 11106012
METB11106009EA 0 CNY
METB11106009 METB11106000 METB11106015 METB11106003 METB11106006 METB11106012
Semi-micro and analytical balances, Excellence XA and XS series
天平
The XS and XA analytical balances provide fast and accurate weighing with flexibility. This is achieved with the introduction of SmartGrid technology, with the gridded weighing pan supported at the rear of the weighing chamber. Therefore the draught shield and drip tray can be completely, quickly and easily removed. The draught shield can be set to open the right hand door with the left slider and vice versa making loading more efficient. The design also features a large backlit touch screen that facilitates direct access to all functions and a standard range of applications, that includeMore Product Information