D-甘露糖醇, EMPROVE® EXPERT USP, BP, Ph. Eur., JP

供应商: MERCK PRODUCTION CHEMICALS

Synonyms: 甘露醇

1.37096.1000
MERP1.37096.1000EA 0 CNY
MERP1.37096.1000
D-甘露糖醇, EMPROVE® EXPERT USP, BP, Ph. Eur., JP
D-甘露糖醇
分子式: HOH₂C(CH(OH))₄CH₂OH
分子量: 182.17 g/mol
沸点: 290…295 °C (4 hPa)
溶点: 164…169 °C
密度: 1.49 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00064287
CAS号: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

现在订购

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR