磷酸二氢钠, anhydrous, LiChropur™ HPLC, Supelco®

供应商: Merck

Synonyms: 磷酸一钠, 磷酸二氢钠

MQ100

5.43840.0250
MERC5.43840.0250EA 0 CNY
MERC5.43840.0250
磷酸二氢钠, anhydrous, LiChropur™ HPLC, Supelco®
磷酸二氢钠
分子式: NaH₂PO₄
分子量: 119.98 g/mol
溶点: 650
存储条件: Ambient
MDL编号: MFCD00003527
CAS号: 7558-80-7
EINECS: 231-449-2
Merck Index: 12,08806

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR