HPLC columns, Superspher®
供应商: MERCK MILLIPORE
1.16857.0001 1.51910.0001
MERC1.16857.0001EA 0 CNY
MERC1.16857.0001 MERC1.51910.0001
HPLC columns, Superspher®
色谱柱
Superspher® improves the separation efficiency of HPLC analysis. This high performance spherical silica carrier, with a mean particle size of 4 μm, yields the best pressure/separation performance ratio in view of today's generation of HPLC systems and according to theoretical calculation and practical experience.
The number of theoretical plates for Superspher® is <100000 N/m. Therefore, they are always first choice if complex mixtures demand high peak capacity.
A broad range of modifications on Superspher® are available: Non polar derivatives (RP-8, RP-8 endcapped, RP-18, RP-18More Product Information