硫酸 0.5 mol/l (1 N) aqueous solution, Titripur® Reag. USP, Reag. Ph. Eur. Volumetric, Supelco®

供应商: Merck
Warning


MQ100

1.09072.1000
MERC1.09072.1000EA 0 CNY
MERC1.09072.1000
硫酸 0.5 mol/l (1 N) aqueous solution, Titripur® Reag. USP, Reag. Ph. Eur. Volumetric, Supelco®
硫酸

Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.


Stability of product in Titripac packaging: Merck have carried out testing to show that products are stable for 36 months when in use and opened. Please see Technical report (Product specification) in the Related Information tab below.

分子式: H₂SO₄
分子量: 98.08 g/mol
沸点: 330
溶点: 10.38
密度: 1.01 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00064589
CAS号: 7664-93-9
EINECS: 231-639-5
UN号: 3264
ADR: 8 III

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR