Potassium chloride saturated with silver chloride aqueous solution 3 mol/l (3 N) aqueous solution Electrodes_filling, Supelco®

供应商: Merck
Warning


MQ100

1.04818.0100
MERC1.04818.0100EA 0 CNY
MERC1.04818.0100
Potassium chloride saturated with silver chloride aqueous solution 3 mol/l (3 N) aqueous solution Electrodes_filling, Supelco®
Potassium chloride saturated with silver chloride aqueous solution
分子式: KCl+AgCl
分子量: 74.55 g/mol
密度: 1.13 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00011360
CAS号: 7447-40-7
EINECS: 231-211-8
UN号: 3082
ADR: 9 III

现在订购

产品技术规格

Solubility (20 °C) soluble
Boiling Point 100 °C (1013 hPa)
Density 1.13 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR