L-组氨酸, Millipore®

供应商: Merck

Synonyms: (S)-2-氨基-3-(4-咪唑基)丙酸

MQ100

1.04351.0100
MERC1.04351.0100EA 0 CNY
MERC1.04351.0100
L-组氨酸, Millipore®
L-组氨酸

Soluble in water and mineral acids; insoluble in organic solvents.

分子式: C₆H₉N₃O₂
分子量: 155.16 g/mol
溶点: 272…273 °C
密度: 1.44 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00064315
CAS号: 71-00-1
EINECS: 200-745-3
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badfdb36aac0611e5823a7d05e6cba944f6650fa&vid=9e9f9d881a5b3314c7f342312e77e13493901200&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR