硫酸铁(II)七水合物, EMSURE® Reag. Ph. Eur., ISO analysis_for, Supelco®

供应商: Merck
Warning

Synonyms: 铁矾

MQ300

1.03965.1000
MERC1.03965.1000EA 0 CNY
MERC1.03965.1000
硫酸铁(II)七水合物, EMSURE® Reag. Ph. Eur., ISO analysis_for, Supelco®
硫酸铁(II)七水合物
分子式: FeSO₄·7H₂O
分子量: 278.02 g/mol
溶点: 64
密度: 1.9 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00149719
CAS号: 7782-63-0
EINECS: 231-753-5
Merck Index: 13,04091

现在订购

产品技术规格

Assay (manganometric) 99.5 - 102.0 %
Identity passes test
Appearance of solution passes test
Insoluble matter ≤ 0.01 %
pH-value (5 %; water) 3.0 - 4.0
Chloride (Cl) ≤ 0.0005 %
Phosphate (PO₄) ≤ 0.001 %
Total nitrogen (N) ≤ 0.001 %
Heavy metals as Pb ≤ 0.005 %
As (Arsenic) ≤ 0.0002 %
Ca (Calcium) ≤ 0.005 %
Cr (Chromium) ≤ 0.0050 %
Cu (Copper) ≤ 0.001 %
Fe III (Iron III) ≤ 0.02 %
K (Potassium) ≤ 0.002 %
Mg (Magnesium) ≤ 0.002 %
Mn (Manganese) ≤ 0.05 %
Na (Sodium) ≤ 0.02 %
Ni (Nickel) ≤ 0.0050 %
Pb (Lead) ≤ 0.0005 %
Zn (Zinc) ≤ 0.0050 %
Co (Cobalt) ≤ 0.0025 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR