二异丙基醚, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®

供应商: Merck
Danger

Synonyms: 异丙醚

MQ300

1.00867.2500
MERC1.00867.2500EA 0 CNY
MERC1.00867.2500
二异丙基醚, EMSURE® Reag. Ph. Eur., ACS analysis_for, Supelco®
二异丙基醚

EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

分子式: (CH₃)₂CHOCH(CH₃)₂
分子量: 102.18 g/mol
沸点: 68.5
溶点: –86 °C
密度: 0.726 g/cm³ (20 °C)
闪点: –28 °C
存储条件: Ambient
MDL编号: MFCD00008880
CAS号: 108-20-3
EINECS: 203-560-6
UN号: 1159
ADR: 3 II
Merck Index: 13,05232
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=932b8c021b2bfa9a11b8d8812cafbafecc6f9239&vid=dea267e876203eb2c2674787701ec602da1b3cf6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Solubility 12 g/l (20 °C)
Melting Point -86 °C
Molar Mass 102.18 g/mol
Boiling Point 67 - 70 °C (1013 hPa)
Vapor Pressure 175 hPa (20 °C)
Flash Point -28 °C
Refractive Index 1.3679 (20 °C, 589 nm)
Explosion Limit 1.0 - 21 %(V)
Density 0.72 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 405 °C
Evaporation number 1.6


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR