Automated pipette tips, Matrix® D.A.R.Ts®
供应商: THERMO MATRIX TECHNOLOGIES
5506 5416 5418 5507 5516
MATR5506EA 0 CNY
MATR5506 MATR5416 MATR5418 MATR5507 MATR5516
Automated pipette tips, Matrix® D.A.R.Ts®
吸头
D.A.R.T’S™ (Disposable Automation Research Tips) have been designed for the ultimate performance in automated pipetting.
  • Small internal orifice provides small droplet formation resulting in optimal accuracy and precision
  • Entire assembly can be quickly removed and replaced eliminating extra handling preventing damage or contamination
  • 100% inspection ensures superior quality
  • Flexibility to choose tips for any application
  • Available in standard, filtered, wide bore and extended length
Unlike conventional automation tips, D.A.R.T.’S are in disposable rigidMore Product Information