Safety cutter, SECUMAX EASYSAFE
供应商: MARTOR ARGENTAX
121001.02
MARO121001.02EA 0 CNY
MARO121001.02
Safety cutter, SECUMAX EASYSAFE
刀和刀具
Steel blade, handle made of glass fibre reinforced plastic.
  • Integrated tape slitter safely cuts packaging tape without an exposed blade
  • Easy blade change - no tools required
  • Cutter can be locked with a special key (must be ordered separately) - avoids the hygiene risk resulting from unauthorised blade changes
  • Blade has four sharp edges and can therefore be rotated four times before it needs to be replaced
  • Ergonomic handle; suitable for left and right-handed users
This plastic and paper sheet cutter features a concealed blade and a cutting head designMore Product Information