Counting chambers with V-slash, Imp. Neubauer
供应商: MARIENFELD
0650010 0650030
MARI0650010EA 0 CNY
MARI0650010 MARI0650030
Counting chambers with V-slash, Imp. Neubauer
载玻片和盖玻片 细胞计数板
These counting chambers with V-slash facilitates the feeding of the capillary gap between chamber bottom and haemacytometer cover glass with reduced risk of overflow.
Certifications: IVD 98/79/EC